Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-CH2CH2-Boc | 1245823-50-0 | 360.44 g·mol⁻¹ | C18H32O7 | 100 MG
SDP

Catalog No. 5000201994
Encompass_Preferred

Propargyl-PEG4-CH2CH2-Boc is a PEG-based linker bearing a terminal propargyl (alkyne) group and a Boc-protected amine. It is used as a non-cleavable ADC linker, a PROTAC linker, and as a reagent for click chemistry (copper-catalyzed azide-alkyne cycloaddition).

  • Contains a terminal alkyne suitable for copper-catalyzed azide-alkyne cycloaddition.
  • Boc-protected amine enables orthogonal deprotection strategies.
  • Applicable as a non-cleavable ADC linker or a PROTAC linker in conjugation workflows.
  • Molecular weight 360.44 g·mol⁻¹; CAS 1245823-50-0.
  • Store pure material at -20°C for long-term stability; in solvent store at -80°C for best stability.

Catalog No. 50-002-01994 Supplier Medchemexpress LLC Supplier No. HY130293100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-CH2CH2-Boc is a PEG-based linker bearing a terminal propargyl (alkyne) group and a Boc-protected amine. It is used as a non-cleavable ADC linker, a PROTAC linker, and as a reagent for click chemistry (copper-catalyzed azide-alkyne cycloaddition).

  • Contains a terminal alkyne suitable for copper-catalyzed azide-alkyne cycloaddition.
  • Boc-protected amine enables orthogonal deprotection strategies.
  • Applicable as a non-cleavable ADC linker or a PROTAC linker in conjugation workflows.
  • Molecular weight 360.44 g·mol⁻¹; CAS 1245823-50-0.
  • Store pure material at -20°C for long-term stability; in solvent store at -80°C for best stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.