Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-CH2CH2-Boc | 1245823-50-0 | MFCD22201534 | 95.0% | 360.44 g/mol | C18H32O7 | 500 MG
SDP

Catalog No. 5000203147
Encompass_Preferred

Propargyl-PEG4-CH2CH2-Boc is a PEG-based linker with a terminal propargyl (alkyne) group and a Boc-protected amine. It is used as a click-chemistry compatible linker for the synthesis of PROTACs and antibody-drug conjugates (ADCs), enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC).

  • Click chemistry compatible (terminal alkyne)
  • Boc-protected amine allows selective deprotection
  • Molecular formula C18H32O7
  • Molecular weight 360.44 g/mol
  • Available in multiple pack sizes, including 500 mg

Catalog No. 50-002-03147 Supplier Medchemexpress LLC Supplier No. HY130293500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-CH2CH2-Boc is a PEG-based linker with a terminal propargyl (alkyne) group and a Boc-protected amine. It is used as a click-chemistry compatible linker for the synthesis of PROTACs and antibody-drug conjugates (ADCs), enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC).

  • Click chemistry compatible (terminal alkyne)
  • Boc-protected amine allows selective deprotection
  • Molecular formula C18H32O7
  • Molecular weight 360.44 g/mol
  • Available in multiple pack sizes, including 500 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.