Learn More
Medchemexpress LLC Propargyl-PEG4-CH2CO2-NHS | 2144777-76-2 | 343.33 | 100 MG
SDP
Catalog No. 5000349203
Encompass_Preferred
Propargyl-PEG4-CH2CO2-NHS (compound P-7) is a PEG derivative containing a propargyl group and an NHS group. It is an ADC linker used in the synthesis of antibody-drug conjugates (ADCs). It is also categorized as a click chemistry reagent, possessing an alkyne group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • PEG derivative with propargyl and NHS groups.
  • Acts as an ADC linker for synthesizing antibody-drug conjugates.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group, allowing for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
Catalog No. 50-003-49203 Supplier Medchemexpress LLC Supplier No. HY140025100MG
Propargyl-PEG4-CH2CO2-NHS (compound P-7) is a PEG derivative containing a propargyl group and an NHS group. It is an ADC linker used in the synthesis of antibody-drug conjugates (ADCs). It is also categorized as a click chemistry reagent, possessing an alkyne group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • PEG derivative with propargyl and NHS groups.
  • Acts as an ADC linker for synthesizing antibody-drug conjugates.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group, allowing for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.