Learn More
Medchemexpress LLC Propargyl-PEG4-CH2CO2-NHS | 2144777-76-2 | 343.33 | 250 MG
SDP
Catalog No. 5000351132
Encompass_Preferred
Propargyl-PEG4-CH2CO2-NHS is a PEG derivative containing both a propargyl group and an NHS group. It functions as an ADC linker, used in the synthesis of antibody-drug conjugates (ADCs). This compound is also a click chemistry reagent, possessing an alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG derivative with propargyl and NHS groups
  • Functions as an ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-51132 Supplier Medchemexpress LLC Supplier No. 250MGHY140025
Propargyl-PEG4-CH2CO2-NHS is a PEG derivative containing both a propargyl group and an NHS group. It functions as an ADC linker, used in the synthesis of antibody-drug conjugates (ADCs). This compound is also a click chemistry reagent, possessing an alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG derivative with propargyl and NHS groups
  • Functions as an ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)