Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-CH2COOH | 1429934-37-1 | ≥97.0% | 290.31 | 1 G
SDP

Catalog No. 5000349164
Encompass_Preferred

Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules. This compound is a colorless to light yellow liquid.

  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
  • Appearance: colorless to light yellow liquid
  • Molecular formula: C13H22O7

Catalog No. 50-003-49164 Supplier Medchemexpress LLC Supplier No. HY1400231G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules. This compound is a colorless to light yellow liquid.

  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
  • Appearance: colorless to light yellow liquid
  • Molecular formula: C13H22O7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.