Learn More
Medchemexpress LLC Propargyl-PEG4-CH2COOH | 1429934-37-1 | >97.0% | 290.31 | 25 MG
SDP
Catalog No. 5000349411
Encompass_Preferred
Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules. PROTACs, composed of two ligands linked together, target an E3 ubiquitin ligase and a specific protein, thereby utilizing the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Features Alkyne group for CuAAc
  • Enables selective protein degradation via ubiquitin-proteasome system
Catalog No. 50-003-49411 Supplier Medchemexpress LLC Supplier No. 25MGHY140023
Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules. PROTACs, composed of two ligands linked together, target an E3 ubiquitin ligase and a specific protein, thereby utilizing the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Features Alkyne group for CuAAc
  • Enables selective protein degradation via ubiquitin-proteasome system