Learn More
Medchemexpress LLC Propargyl-PEG4-CH2COOH | 1429934-37-1 | ≥97.0% | 290.31 | 250 MG
SDP
Catalog No. 5000348790
Encompass_Preferred
Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker that is utilized in the synthesis of PROTACs. This compound also serves as a click chemistry reagent, featuring an Alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-48790 Supplier Medchemexpress LLC Supplier No. 250MGHY140023
Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker that is utilized in the synthesis of PROTACs. This compound also serves as a click chemistry reagent, featuring an Alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)