Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-GGFG-DXd | 2762518-94-3 | 100.0% | 1083.12 g/mol | C54H63FN8O15 | 5 MG
SDP

Catalog No. 5000204362
Encompass_Preferred

Propargyl-PEG4-GGFG-DXd is a drug-linker conjugate for antibody-drug conjugate (ADC) research that couples a cleavable GGFG peptide-PEG4 linker with a propargyl handle to the DXd topoisomerase I inhibitor. It is supplied as an off-white to light yellow solid, with high purity and DMSO solubility for use in conjugation and payload studies.

  • High purity, 99.99% (HPLC)
  • Molecular weight 1083.12 g/mol
  • Chemical formula C54H63FN8O15
  • Physical appearance: solid, off-white to light yellow
  • Storage: 4°C as supplied; in solvent -80°C (up to 6 months) or -20°C (up to 1 month)
  • Solubility: DMSO up to 100 mg/mL (use ultrasonic)
  • Intended use: research use only for ADC linker-payload development

Catalog No. 50-002-04362 Supplier Medchemexpress LLC Supplier No. HY1567555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-GGFG-DXd is a drug-linker conjugate for antibody-drug conjugate (ADC) research that couples a cleavable GGFG peptide-PEG4 linker with a propargyl handle to the DXd topoisomerase I inhibitor. It is supplied as an off-white to light yellow solid, with high purity and DMSO solubility for use in conjugation and payload studies.

  • High purity, 99.99% (HPLC)
  • Molecular weight 1083.12 g/mol
  • Chemical formula C54H63FN8O15
  • Physical appearance: solid, off-white to light yellow
  • Storage: 4°C as supplied; in solvent -80°C (up to 6 months) or -20°C (up to 1 month)
  • Solubility: DMSO up to 100 mg/mL (use ultrasonic)
  • Intended use: research use only for ADC linker-payload development

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.