Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-GGFG-DXd | 2762518-94-3 | MFCD01346226 | 100.0% | 1083.12 | C54H63FN8O15 | 10 MG
SDP

Catalog No. 5000204396
Encompass_Preferred

Propargyl-PEG4-GGFG-DXd is a pre-formed drug-linker conjugate intended for research use in antibody-drug conjugate (ADC) development. It combines a PEG4-linked GGFG tetrapeptide linker with a DXd topoisomerase I inhibitor payload to support linker evaluation and conjugation workflows.

  • Contains a DXd topoisomerase I inhibitor payload.
  • Includes a PEG4 spacer and GGFG tetrapeptide linker for conjugation.
  • Off-white to light yellow solid, high purity suitable for analytical work.
  • Soluble in DMSO at up to 100 mg/mL with ultrasonic assistance.
  • Stable when stored sealed and protected from moisture; in solvent: -80 °C (6 months), -20 °C (1 month).

Catalog No. 50-002-04396 Supplier Medchemexpress LLC Supplier No. HY15675510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-GGFG-DXd is a pre-formed drug-linker conjugate intended for research use in antibody-drug conjugate (ADC) development. It combines a PEG4-linked GGFG tetrapeptide linker with a DXd topoisomerase I inhibitor payload to support linker evaluation and conjugation workflows.

  • Contains a DXd topoisomerase I inhibitor payload.
  • Includes a PEG4 spacer and GGFG tetrapeptide linker for conjugation.
  • Off-white to light yellow solid, high purity suitable for analytical work.
  • Soluble in DMSO at up to 100 mg/mL with ultrasonic assistance.
  • Stable when stored sealed and protected from moisture; in solvent: -80 °C (6 months), -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.