Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-GGFG-DXd | 2762518-94-3 | MFCD01346226 | 100.0% | 1083.12 | C54H63FN8O15 | 100 MG
SDP

Catalog No. 5000204463
Encompass_Preferred

Propargyl-PEG4-GGFG-DXd is a drug-linker conjugate for antibody-drug conjugates (ADCs) that links an ADC linker to the DNA topoisomerase I inhibitor DXd and provides a propargyl handle for bioorthogonal conjugation.

  • Molecular formula: C54H63FN8O15
  • Molecular weight: 1083.12
  • Cas number: 2762518-94-3
  • Purity: 99.99%
  • Appearance: solid, off-white to light yellow
  • Storage: 4°C sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month
  • Package size: available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg; larger sizes available by quote

Catalog No. 50-002-04463 Supplier Medchemexpress LLC Supplier No. HY156755100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-GGFG-DXd is a drug-linker conjugate for antibody-drug conjugates (ADCs) that links an ADC linker to the DNA topoisomerase I inhibitor DXd and provides a propargyl handle for bioorthogonal conjugation.

  • Molecular formula: C54H63FN8O15
  • Molecular weight: 1083.12
  • Cas number: 2762518-94-3
  • Purity: 99.99%
  • Appearance: solid, off-white to light yellow
  • Storage: 4°C sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month
  • Package size: available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg; larger sizes available by quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.