Learn More
Medchemexpress LLC Propargyl-PEG4-NHS ester | 1428629-70-2 | 97.0% | 357.36 g·mol⁻¹ | C16H23NO8 | 100 MG
SDP
Catalog No. 5000201889
Encompass_Preferred
Propargyl-PEG4-NHS ester is a PEG4 linker with a terminal propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) ester, designed for bioconjugation via amine coupling and click chemistry.
  • Noncleavable four-unit PEG linker for spacer and solubility
  • Terminal propargyl group enables copper-catalyzed azide-alkyne cycloaddition
  • NHS ester provides rapid and efficient coupling to primary amines
  • High purity suitable for research applications
  • Molecular weight approximately 357.36 g·mol⁻¹
  • Available in small research-scale package sizes
Catalog No. 50-002-01889 Supplier Medchemexpress LLC Supplier No. HY126516100MG
Propargyl-PEG4-NHS ester is a PEG4 linker with a terminal propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) ester, designed for bioconjugation via amine coupling and click chemistry.
  • Noncleavable four-unit PEG linker for spacer and solubility
  • Terminal propargyl group enables copper-catalyzed azide-alkyne cycloaddition
  • NHS ester provides rapid and efficient coupling to primary amines
  • High purity suitable for research applications
  • Molecular weight approximately 357.36 g·mol⁻¹
  • Available in small research-scale package sizes