Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-NHS ester | 1428629-70-2 | MFCD22380743 | 97.0% | 357.36 g/mol | C16H23NO8 | 50 MG
SDP

Catalog No. 5000201905
Encompass_Preferred

Propargyl-PEG4-NHS ester is a propargyl-terminated polyethylene glycol (PEG4) N-hydroxysuccinimide (NHS) ester used as a linker and click-chemistry reagent for bioconjugation and small-molecule attachment. It combines a reactive NHS ester for amine coupling with an alkyne handle for copper-catalyzed azide-alkyne cycloaddition.

  • Provides an alkyne handle for click chemistry.
  • Contains an NHS ester for efficient amine coupling.
  • Four-unit PEG spacer improves solubility and flexibility.
  • Suitable for linker construction, ADC and PROTAC assembly, and general bioconjugation.
  • Storage: typically kept frozen to preserve reactivity.

Catalog No. 50-002-01905 Supplier Medchemexpress LLC Supplier No. HY12651650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-NHS ester is a propargyl-terminated polyethylene glycol (PEG4) N-hydroxysuccinimide (NHS) ester used as a linker and click-chemistry reagent for bioconjugation and small-molecule attachment. It combines a reactive NHS ester for amine coupling with an alkyne handle for copper-catalyzed azide-alkyne cycloaddition.

  • Provides an alkyne handle for click chemistry.
  • Contains an NHS ester for efficient amine coupling.
  • Four-unit PEG spacer improves solubility and flexibility.
  • Suitable for linker construction, ADC and PROTAC assembly, and general bioconjugation.
  • Storage: typically kept frozen to preserve reactivity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.