Learn More
Medchemexpress LLC Propargyl-PEG4-O-C1-Boc | 2098489-63-3 | 97% | 100 MG
SDP
Catalog No. 5000341021
Encompass_Preferred
Propargyl-PEG4-O-C1-Boc is a versatile PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This compound also functions as a click chemistry reagent, featuring an Alkyne group that enables efficient copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide groups
Catalog No. 50-003-41021 Supplier Medchemexpress LLC Supplier No. HY130389100MG
Propargyl-PEG4-O-C1-Boc is a versatile PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This compound also functions as a click chemistry reagent, featuring an Alkyne group that enables efficient copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide groups