Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-O-C1-Boc | 2098489-63-3 | 97% | 250 MG
SDP

Catalog No. 5000341791
Encompass_Preferred

Propargyl-PEG4-O-C1-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a versatile click chemistry reagent, featuring an Alkyne group that readily participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing Azide groups.

  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Click chemistry reagent with Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Molecular formula: C17H30O7
  • Molecular weight: 346.42
  • For research use only

Catalog No. 50-003-41791 Supplier Medchemexpress LLC Supplier No. 250MGHY130389
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-O-C1-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a versatile click chemistry reagent, featuring an Alkyne group that readily participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing Azide groups.

  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Click chemistry reagent with Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Molecular formula: C17H30O7
  • Molecular weight: 346.42
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.