Learn More
Medchemexpress LLC Propargyl-PEG4-O-C1-NHS ester | 1161883-51-7 | 25 MG
SDP
Catalog No. 5000342780
Encompass_Preferred
Propargyl-PEG4-O-C1-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Enables selective degradation of target proteins
Catalog No. 50-003-42780 Supplier Medchemexpress LLC Supplier No. 25MGHY130390
Propargyl-PEG4-O-C1-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Enables selective degradation of target proteins