Learn More
Medchemexpress LLC Propargyl-PEG4-O-C1-NHS ester | 1161883-51-7 | 500 MG
SDP
Catalog No. 5000340645
Encompass_Preferred
This compound is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-40645 Supplier Medchemexpress LLC Supplier No. HY130390500MG
This compound is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)