Learn More
Medchemexpress LLC Propargyl-PEG4-sulfonic acid | 1817735-29-7 | 98.0% | 296.34 | 5 G
SDP
Catalog No. 5000350303
Encompass_Preferred
Propargyl-PEG4-sulfonic acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-50303 Supplier Medchemexpress LLC Supplier No. HY1400475G
Propargyl-PEG4-sulfonic acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc)