Learn More
Medchemexpress LLC Propargyl-PEG4-Tos | 875770-32-4 | 99.3% | 5 G
SDP
Catalog No. 5000340449
Encompass_Preferred
Propargyl-PEG4-Tos is a PEG-based PROTAC linker used in the synthesis of PROTACs and as a cleavable ADC linker in the synthesis of antibody-drug conjugates (ADCs). It also functions as a click chemistry reagent featuring an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Cleavable ADC linker
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)
  • Click chemistry reagent
  • Features an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
Catalog No. 50-003-40449 Supplier Medchemexpress LLC Supplier No. HY1303875G
Propargyl-PEG4-Tos is a PEG-based PROTAC linker used in the synthesis of PROTACs and as a cleavable ADC linker in the synthesis of antibody-drug conjugates (ADCs). It also functions as a click chemistry reagent featuring an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Cleavable ADC linker
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)
  • Click chemistry reagent
  • Features an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules