Learn More
Medchemexpress LLC Propargyl-PEG4-Tos | 875770-32-4 | 99.3% | 500 MG
SDP
Catalog No. 5000338328
Encompass_Preferred
Propargyl-PEG4-Tos is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a cleavable ADC linker for synthesizing antibody-drug conjugates (ADCs) and serves as a click chemistry reagent containing an Alkyne group. This allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Cleavable ADC linker
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-38328 Supplier Medchemexpress LLC Supplier No. HY130387500MG
Propargyl-PEG4-Tos is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a cleavable ADC linker for synthesizing antibody-drug conjugates (ADCs) and serves as a click chemistry reagent containing an Alkyne group. This allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Cleavable ADC linker
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)