Learn More
Medchemexpress LLC Propargyl-PEG5-amine | 1589522-46-2 | MFCD27977505 | 98.9% | 275.34 g/mol | C13H25NO5 | 250 MG
SDP
Catalog No. 5000204194
Encompass_Preferred
Propargyl-PEG5-amine is a PEG-based linker reagent used as an alkyne functional handle for click chemistry and as a non-cleavable linker in ADC and PROTAC synthesis.
  • PEG-based alkyne linker for conjugation and click chemistry.
  • Non-cleavable linker suitable for ADC and PROTAC applications.
  • Provides an alkyne handle for copper-catalyzed azide-alkyne cycloaddition.
  • High purity and fully characterized by LCMS, HNMR, and RP-HPLC.
  • Available in multiple pack sizes, including 250 MG.
  • Colorless to light yellow liquid with a density of 1.049 g/cm3.
Catalog No. 50-002-04194 Supplier Medchemexpress LLC Supplier No. HY126976250MG
Propargyl-PEG5-amine is a PEG-based linker reagent used as an alkyne functional handle for click chemistry and as a non-cleavable linker in ADC and PROTAC synthesis.
  • PEG-based alkyne linker for conjugation and click chemistry.
  • Non-cleavable linker suitable for ADC and PROTAC applications.
  • Provides an alkyne handle for copper-catalyzed azide-alkyne cycloaddition.
  • High purity and fully characterized by LCMS, HNMR, and RP-HPLC.
  • Available in multiple pack sizes, including 250 MG.
  • Colorless to light yellow liquid with a density of 1.049 g/cm3.