Learn More
Medchemexpress LLC Propargyl-PEG5-Br | 1287660-83-6 | 339.22 | 1 G
SDP
Catalog No. 5000351973
Encompass_Preferred
Propargyl-PEG5-Br is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs and functions as a click chemistry reagent. It contains an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Enables synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Features an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
  • Supports development of PROTACs for target protein degradation
Catalog No. 50-003-51973 Supplier Medchemexpress LLC Supplier No. HY1387361G
Propargyl-PEG5-Br is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs and functions as a click chemistry reagent. It contains an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Enables synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Features an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
  • Supports development of PROTACs for target protein degradation