Learn More
Medchemexpress LLC Propargyl-PEG5-Br | 1287660-83-6 | 339.22 | 100 MG
SDP
Catalog No. 5000350056
Encompass_Preferred
Propargyl-PEG5-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50056 Supplier Medchemexpress LLC Supplier No. HY138736100MG
Propargyl-PEG5-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins