Learn More
Medchemexpress LLC Propargyl-PEG5-Br | 1287660-83-6 | 339.22 | 250 MG
SDP
Catalog No. 5000347835
Encompass_Preferred
Propargyl-PEG5-Br is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This linker is ideal for researchers developing novel protein degradation therapeutics.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-47835 Supplier Medchemexpress LLC Supplier No. 250MGHY138736
Propargyl-PEG5-Br is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This linker is ideal for researchers developing novel protein degradation therapeutics.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups