Learn More
Medchemexpress LLC Propargyl-PEG5-NHS ester | 1393330-40-9 | 98.0% | 1 G
SDP
Catalog No. 5000340911
Encompass_Preferred
Propargyl-PEG5-NHS ester is a PEG/Alkyl/ether-based PROTAC linker primarily used in the synthesis of PROTACs. It also functions as a cleavable ADC linker, crucial for synthesizing antibody-drug conjugates (ADCs).
  • Functions as a cleavable ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Click chemistry reagent
  • Contains an alkyne group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-40911 Supplier Medchemexpress LLC Supplier No. HY1303881G
Propargyl-PEG5-NHS ester is a PEG/Alkyl/ether-based PROTAC linker primarily used in the synthesis of PROTACs. It also functions as a cleavable ADC linker, crucial for synthesizing antibody-drug conjugates (ADCs).
  • Functions as a cleavable ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Click chemistry reagent
  • Contains an alkyne group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups