Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargyl-PEG5-NHS Ester, TCI America™
SDP

Catalog No. P228325MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P228325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P228325MG Supplier TCI America Supplier No. P228325MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1393330-40-9
Molecular Formula C18H27NO9
Molecular Weight (g/mol) 401.412
MDL Number MFCD22574798
InChI Key ACZDUKRWTPZVRP-UHFFFAOYSA-N
Synonym Alkyne-PEG5-NHS Ester, N-Succinimidyl 4,7,10,13,16-Pentaoxanonadec-18-ynoate, 4,7,10,13,16-Pentaoxanonadec-18-ynoic Acid N-Succinimidyl Ester
PubChem CID 60146234
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethoxy]propanoate
SMILES C#CCOCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O

Specifications

Quantity 25 mg
Formula Weight 401.41
Physical Form Liquid
Chemical Name or Material Propargyl-PEG5-NHS Ester
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.