Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG6-acid | 1951438-84-8 | 348.39 g·mol⁻1 | C16H28O8 | 100 MG
SDP

Catalog No. 5000202720
Encompass_Preferred

Propargyl-PEG6-acid is a PEG-based linker bearing a terminal propargyl (alkyne) group, used as a click-chemistry reagent and as a linker in the synthesis of PROTACs and antibody-drug conjugates. The PEG6 spacer increases hydrophilicity and enables conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC).

  • Contains a terminal propargyl (alkyne) functional group.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Used in synthesis of PROTACs and antibody-drug conjugates.
  • PEG6 spacer provides hydrophilicity and improved solubility.
  • Storage recommendations: pure form refrigerated or frozen for long-term stability; store solutions frozen.

Catalog No. 50-002-02720 Supplier Medchemexpress LLC Supplier No. HY130386100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG6-acid is a PEG-based linker bearing a terminal propargyl (alkyne) group, used as a click-chemistry reagent and as a linker in the synthesis of PROTACs and antibody-drug conjugates. The PEG6 spacer increases hydrophilicity and enables conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC).

  • Contains a terminal propargyl (alkyne) functional group.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Used in synthesis of PROTACs and antibody-drug conjugates.
  • PEG6 spacer provides hydrophilicity and improved solubility.
  • Storage recommendations: pure form refrigerated or frozen for long-term stability; store solutions frozen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.