Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-peg6-acid | 1951438-84-8 | 348.39 g/mol | C16H28O8 | 50 MG
SDP

Catalog No. 5000204135
Encompass_Preferred

Propargyl-PEG6-acid is a PEG6-based linker and click-chemistry reagent containing a terminal propargyl (alkyne) group and a carboxylic acid. It is used to introduce a PEG6 spacer in the synthesis of PROTACs and antibody-drug conjugates (ADCs) and undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation.

  • Used as PEG6 spacer in PROTAC and ADC synthesis.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Molecular weight 348.39 g/mol; formula C16H28O8.
  • CAS number 1951438-84-8 for unambiguous identification.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-04135 Supplier Medchemexpress LLC Supplier No. HY13038650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG6-acid is a PEG6-based linker and click-chemistry reagent containing a terminal propargyl (alkyne) group and a carboxylic acid. It is used to introduce a PEG6 spacer in the synthesis of PROTACs and antibody-drug conjugates (ADCs) and undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation.

  • Used as PEG6 spacer in PROTAC and ADC synthesis.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Molecular weight 348.39 g/mol; formula C16H28O8.
  • CAS number 1951438-84-8 for unambiguous identification.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.