Learn More
Medchemexpress LLC Propargyl-PEG6-acid | 1951438-84-8 | 5 G
SDP
Catalog No. 5000338327
Encompass_Preferred
Propargyl-PEG6-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). This compound is a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing Azide groups.
  • Peg-based protac linker for protac synthesis.
  • Cleavable adc linker for antibody-drug conjugate synthesis.
  • Click chemistry reagent containing an alkyne group.
  • Enables copper-catalyzed azide-alkyne cycloaddition (cuaac) with azide groups.
Catalog No. 50-003-38327 Supplier Medchemexpress LLC Supplier No. HY1303865G
Propargyl-PEG6-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). This compound is a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing Azide groups.
  • Peg-based protac linker for protac synthesis.
  • Cleavable adc linker for antibody-drug conjugate synthesis.
  • Click chemistry reagent containing an alkyne group.
  • Enables copper-catalyzed azide-alkyne cycloaddition (cuaac) with azide groups.