Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG6-acid | 1951438-84-8 | ≥97.0% | 348.39 g/mol | C16H28O8 | 250 MG
SDP

Catalog No. 5000202503
Encompass_Preferred

Propargyl-PEG6-acid is a PEG-based linker and click chemistry reagent containing a terminal alkyne. It is used as a linker in PROTAC and ADC synthesis and for copper-catalyzed azide-alkyne cycloaddition (CuAAc) bioconjugation reactions.

  • cleavable PEG6 linker suitable for PROTAC and ADC synthesis
  • contains a terminal propargyl (alkyne) group for click chemistry
  • high purity (≥97.0% by NMR) suitable for research applications
  • molecular weight 348.39 g/mol, molecular formula C16H28O8
  • storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-02503 Supplier Medchemexpress LLC Supplier No. HY130386250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG6-acid is a PEG-based linker and click chemistry reagent containing a terminal alkyne. It is used as a linker in PROTAC and ADC synthesis and for copper-catalyzed azide-alkyne cycloaddition (CuAAc) bioconjugation reactions.

  • cleavable PEG6 linker suitable for PROTAC and ADC synthesis
  • contains a terminal propargyl (alkyne) group for click chemistry
  • high purity (≥97.0% by NMR) suitable for research applications
  • molecular weight 348.39 g/mol, molecular formula C16H28O8
  • storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.