Learn More
Medchemexpress LLC Propargyl-PEG6-acid | 1951438-84-8 | MFCD28950778 | >98.0% | 348.39 | C16H28O8 | 1 G
SDP
Catalog No. 5000203915
Encompass_Preferred
Propargyl-PEG6-acid is a polyethylene glycol (PEG6) linker bearing a terminal propargyl (alkyne) group and a carboxylic acid functional handle, used for click chemistry conjugation such as copper-catalyzed azide-alkyne cycloaddition (CuAAC) in ADC, PROTAC, and bioconjugation workflows.
  • Terminal alkyne functional handle for CuAAC click reactions.
  • Carboxylic acid group enables amide coupling and further derivatization.
  • PEG6 spacer imparts hydrophilicity and flexible linker properties.
  • Molecular weight approximately 348.39, suitable for small-molecule linker applications.
  • Available in common laboratory quantities for synthesis and conjugation workflows.
Catalog No. 50-002-03915 Supplier Medchemexpress LLC Supplier No. HY1303861G
Propargyl-PEG6-acid is a polyethylene glycol (PEG6) linker bearing a terminal propargyl (alkyne) group and a carboxylic acid functional handle, used for click chemistry conjugation such as copper-catalyzed azide-alkyne cycloaddition (CuAAC) in ADC, PROTAC, and bioconjugation workflows.
  • Terminal alkyne functional handle for CuAAC click reactions.
  • Carboxylic acid group enables amide coupling and further derivatization.
  • PEG6 spacer imparts hydrophilicity and flexible linker properties.
  • Molecular weight approximately 348.39, suitable for small-molecule linker applications.
  • Available in common laboratory quantities for synthesis and conjugation workflows.