Learn More
Medchemexpress LLC Propargyl-PEG6-alcohol | 944560-99-0 | 97.0% | 1 G
SDP
Catalog No. 5000340450
Encompass_Preferred
Propargyl-PEG6-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used in the synthesis of proteolysis targeting chimeras (PROTACs)
  • Contains an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-40450 Supplier Medchemexpress LLC Supplier No. HY1303821G
Propargyl-PEG6-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used in the synthesis of proteolysis targeting chimeras (PROTACs)
  • Contains an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins