Learn More
Medchemexpress LLC Propargyl-PEG6-alcohol | 944560-99-0 | 97.0% | 250 MG
SDP
Catalog No. 5000341556
Encompass_Preferred
Propargyl-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This makes it suitable for various chemical synthesis applications.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Used in the synthesis of PROTACs
Catalog No. 50-003-41556 Supplier Medchemexpress LLC Supplier No. 250MGHY130382
Propargyl-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This makes it suitable for various chemical synthesis applications.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Used in the synthesis of PROTACs