Learn More
Medchemexpress LLC Propargyl-PEG6-N3 | 1198080-03-3 | 100 MG
SDP
Catalog No. 5000339617
Encompass_Preferred
Propargyl-PEG6-N3 is a PEG-based PROTAC linker employed in the synthesis of PROTACs. It functions as a versatile click chemistry reagent, featuring an Azide group. This group readily participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) when combined with molecules containing Alkyne groups. Furthermore, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules that possess DBCO or BCN groups, offering flexible synthetic pathways.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Click chemistry reagent.
  • Contains an Azide group.
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
Catalog No. 50-003-39617 Supplier Medchemexpress LLC Supplier No. HY130183100MG
Propargyl-PEG6-N3 is a PEG-based PROTAC linker employed in the synthesis of PROTACs. It functions as a versatile click chemistry reagent, featuring an Azide group. This group readily participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) when combined with molecules containing Alkyne groups. Furthermore, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules that possess DBCO or BCN groups, offering flexible synthetic pathways.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Click chemistry reagent.
  • Contains an Azide group.
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.