Learn More
Medchemexpress LLC Propargyl-PEG6-N3 | 1198080-03-3 | 25 MG
SDP
Catalog No. 5000338795
Encompass_Preferred
Propargyl-PEG6-N3 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Engages in strain-promoted alkyne-azide cycloaddition reactions
Catalog No. 50-003-38795 Supplier Medchemexpress LLC Supplier No. 25MGHY130183
Propargyl-PEG6-N3 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Engages in strain-promoted alkyne-azide cycloaddition reactions