Learn More
Medchemexpress LLC Propargyl-PEG6-N3 | 1198080-03-3 | MFCD31561110 | >98.0% | C15H27N3O6 | 10MG
SDP
Catalog No. 5000259782
Encompass_Preferred
Propargyl-PEG6-N3 is a PEG-based linker that contains an azide and a terminal propargyl (alkyne) group, designed for click chemistry and bioconjugation. It is used as a spacer in PROTAC synthesis and in the modular assembly of labeled biomolecules, and should be stored per supplier recommendations.
Catalog No. 50-002-59782 Supplier Medchemexpress LLC Supplier No. HY13018310MG
Propargyl-PEG6-N3 is a PEG-based linker that contains an azide and a terminal propargyl (alkyne) group, designed for click chemistry and bioconjugation. It is used as a spacer in PROTAC synthesis and in the modular assembly of labeled biomolecules, and should be stored per supplier recommendations.