Learn More
Medchemexpress LLC Propargyl-PEG6-NH2 | 1198080-04-4 | 99.7% | 250 MG
SDP
Catalog No. 5000342000
Encompass_Preferred
Propargyl-PEG6-NH2 is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Exploits the intracellular ubiquitin-proteasome system to degrade target proteins
Catalog No. 50-003-42000 Supplier Medchemexpress LLC Supplier No. 250MGHY130180
Propargyl-PEG6-NH2 is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Exploits the intracellular ubiquitin-proteasome system to degrade target proteins