Learn More
Medchemexpress LLC Propargyl-PEG6-NH2 | 1198080-04-4 | 99.7% | 5 G
SDP
Catalog No. 5000340069
Encompass_Preferred
Propargyl-PEG6-NH2 is a PEG-based PROTAC linker that functions as a click chemistry reagent. It contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. This reagent is primarily used in the synthesis of PROTACs, which leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules possessing Azide groups
Catalog No. 50-003-40069 Supplier Medchemexpress LLC Supplier No. HY1301805G
Propargyl-PEG6-NH2 is a PEG-based PROTAC linker that functions as a click chemistry reagent. It contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. This reagent is primarily used in the synthesis of PROTACs, which leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules possessing Azide groups