Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG6-NHS ester | 2093153-99-0 | 98.0% | 445.46 g·mol^-1 | C20H31NO10 | 500 MG
SDP

Supplier:  Medchemexpress LLC HY130385500MG

Encompass_Preferred

Propargyl-PEG6-NHS ester is an alkyne-functionalized polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester for amine-reactive conjugation. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and amide bond formation for bioconjugation applications such as PROTAC and ADC synthesis.

  • Contains a terminal propargyl (alkyne) group for click chemistry.
  • Contains an NHS ester for efficient conjugation to primary amines.
  • PEG6 spacer provides flexibility and aqueous solubility.
  • Typical purity 98.0% as supplied.
  • Appearance: colorless to light yellow liquid.
  • Store sealed, away from moisture and light at -20°C; in solvent store at -80°C for long-term.

Catalog No. 50-002-03378


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.