Learn More
Medchemexpress LLC Propargyl-PEG7-acid | 2093154-00-6 | ≥98.0% | 1 G
SDP
Catalog No. 5000338405
Encompass_Preferred
Propargyl-PEG7-acid is a PEG-based PROTAC linker and a cleavable ADC linker, making it valuable for synthesizing PROTACs and antibody-drug conjugates. It also functions as a click chemistry reagent, enabling copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Used in the synthesis of PROTACs
  • Cleavable ADC linker
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition
Catalog No. 50-003-38405 Supplier Medchemexpress LLC Supplier No. HY1303831G
Propargyl-PEG7-acid is a PEG-based PROTAC linker and a cleavable ADC linker, making it valuable for synthesizing PROTACs and antibody-drug conjugates. It also functions as a click chemistry reagent, enabling copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Used in the synthesis of PROTACs
  • Cleavable ADC linker
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition