Learn More
Medchemexpress LLC Propargyl-PEG7-Boc | 1818294-29-9 | 100 MG
SDP
Catalog No. 5000338404
Encompass_Preferred
Propargyl-PEG7-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules. PROTACs are compounds composed of two distinct ligands joined by a linker; one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is for research purposes only and not for human use.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Enables selective degradation of target proteins
Catalog No. 50-003-38404 Supplier Medchemexpress LLC Supplier No. HY130380100MG
Propargyl-PEG7-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules. PROTACs are compounds composed of two distinct ligands joined by a linker; one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is for research purposes only and not for human use.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Enables selective degradation of target proteins