Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG7-Boc | 1818294-29-9 | >95% | 1 G
SDP

Catalog No. 5000340914
Encompass_Preferred

Propargyl-PEG7-Boc is a polyethylene glycol (PEG)-based PROTAC linker. It can be used in the synthesis of a series of PROTACs.

  • Polyethylene glycol (PEG)-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity >95%
  • Soluble in DCM, DMF, DMSO
  • Appearance: Pale yellow or colorless oily liquid

Catalog No. 50-003-40914 Supplier Medchemexpress LLC Supplier No. HY1303801G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG7-Boc is a polyethylene glycol (PEG)-based PROTAC linker. It can be used in the synthesis of a series of PROTACs.

  • Polyethylene glycol (PEG)-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity >95%
  • Soluble in DCM, DMF, DMSO
  • Appearance: Pale yellow or colorless oily liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.