Learn More
Medchemexpress LLC Propargyl-PEG7-NHS ester | 2093152-77-1 | MFCD29041936 | >95.0% | 489.51 g/mol | C22H35NO11 | 1 G
SDP
Catalog No. 5000204027
Encompass_Preferred
Propargyl-PEG7-NHS ester is an alkyne-functional PEG7 N-hydroxysuccinimide (NHS) ester used as a bifunctional linker for bioconjugation. It couples to primary amines via NHS chemistry and provides a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition, making it suitable for ADC, PROTAC, and general labeling applications.
  • Alkyne-functional terminal enables click chemistry.
  • NHS ester reacts with primary amines for covalent coupling.
  • PEG7 spacer increases solubility and provides flexibility.
  • Molecular weight about 489.51 g/mol and formula C22H35NO11.
  • Typical applications include ADC linkers, PROTAC synthesis, and peptide or protein labeling.
Catalog No. 50-002-04027 Supplier Medchemexpress LLC Supplier No. HY1303811G
Propargyl-PEG7-NHS ester is an alkyne-functional PEG7 N-hydroxysuccinimide (NHS) ester used as a bifunctional linker for bioconjugation. It couples to primary amines via NHS chemistry and provides a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition, making it suitable for ADC, PROTAC, and general labeling applications.
  • Alkyne-functional terminal enables click chemistry.
  • NHS ester reacts with primary amines for covalent coupling.
  • PEG7 spacer increases solubility and provides flexibility.
  • Molecular weight about 489.51 g/mol and formula C22H35NO11.
  • Typical applications include ADC linkers, PROTAC synthesis, and peptide or protein labeling.