Learn More
Medchemexpress LLC Propargyl-PEG8-acid | 2055014-94-1 | 98.0% | 436.49 | C20H36O10 | 250 MG
SDP
Catalog No. 5000203376
Encompass_Preferred
Propargyl-PEG8-acid is an alkyne-functionalized PEG8 linker used as a cleavable linker in PROTAC and antibody-drug conjugate synthesis. It functions as a click chemistry reagent (terminal alkyne) for copper-catalyzed azide-alkyne cycloaddition and is supplied with high purity for medicinal chemistry applications.
  • Alkyne-terminated PEG8 linker suitable for CuAAC click reactions.
  • Cleavable linker for PROTAC and antibody-drug conjugate (ADC) assembly.
  • Molecular formula C20H36O10; molecular weight 436.49.
  • High purity laboratory grade, suitable for synthesis.
  • Available in multiple pack sizes including 250 mg.
Catalog No. 50-002-03376 Supplier Medchemexpress LLC Supplier No. HY130379250MG
Propargyl-PEG8-acid is an alkyne-functionalized PEG8 linker used as a cleavable linker in PROTAC and antibody-drug conjugate synthesis. It functions as a click chemistry reagent (terminal alkyne) for copper-catalyzed azide-alkyne cycloaddition and is supplied with high purity for medicinal chemistry applications.
  • Alkyne-terminated PEG8 linker suitable for CuAAC click reactions.
  • Cleavable linker for PROTAC and antibody-drug conjugate (ADC) assembly.
  • Molecular formula C20H36O10; molecular weight 436.49.
  • High purity laboratory grade, suitable for synthesis.
  • Available in multiple pack sizes including 250 mg.