Learn More
Medchemexpress LLC Propargyl-PEG8-acid | 2055014-94-1 | 98.0% | 436.49 g/mol | C20H36O10 | 1 G
SDP
Catalog No. 5000203374
Encompass_Preferred
Propargyl-PEG8-acid is a polyethylene glycol (PEG) based linker bearing a terminal propargyl (alkyne) group and a carboxylic acid, used for conjugation in PROTAC and antibody-drug conjugate (ADC) synthesis.
  • Contains a terminal propargyl alkyne for click chemistry.
  • Provides a carboxylic acid handle for amide coupling.
  • Approximate molecular weight 436.49 g/mol.
  • High purity (98.0%) suitable for research applications.
  • Available in multiple pack sizes, including 1 G for lab-scale use.
Catalog No. 50-002-03374 Supplier Medchemexpress LLC Supplier No. HY1303791G
Propargyl-PEG8-acid is a polyethylene glycol (PEG) based linker bearing a terminal propargyl (alkyne) group and a carboxylic acid, used for conjugation in PROTAC and antibody-drug conjugate (ADC) synthesis.
  • Contains a terminal propargyl alkyne for click chemistry.
  • Provides a carboxylic acid handle for amide coupling.
  • Approximate molecular weight 436.49 g/mol.
  • High purity (98.0%) suitable for research applications.
  • Available in multiple pack sizes, including 1 G for lab-scale use.