Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG8-acid | 2055014-94-1 | 98.0% | 436.49 g/mol | C20H36O10 | 100 MG
SDP

Catalog No. 5000202716
Encompass_Preferred

Propargyl-PEG8-acid is a propargyl-functionalized polyethylene glycol (PEG8) carboxylic acid linker designed for bioconjugation and linker chemistry. It provides a PEG8 spacer with a terminal alkyne for click chemistry and a carboxylic acid for coupling reactions; it is supplied as a high-purity solid with defined stability and storage requirements.

  • Propargyl terminal alkyne suitable for copper-catalyzed click chemistry.
  • Peg8 spacer improves solubility and flexibility in conjugates.
  • Carboxylic acid functionality enables amide coupling and derivatization.
  • High purity (approximately 98.0%) for reliable synthetic performance.
  • Molecular weight 436.49 g/mol, formula C20H36O10.
  • Available in small-scale packs (100 mg) and larger sizes for scale-up.
  • Store at -20°C under inert atmosphere; in solvent, follow recommended cold storage times.

Catalog No. 50-002-02716 Supplier Medchemexpress LLC Supplier No. HY130379100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG8-acid is a propargyl-functionalized polyethylene glycol (PEG8) carboxylic acid linker designed for bioconjugation and linker chemistry. It provides a PEG8 spacer with a terminal alkyne for click chemistry and a carboxylic acid for coupling reactions; it is supplied as a high-purity solid with defined stability and storage requirements.

  • Propargyl terminal alkyne suitable for copper-catalyzed click chemistry.
  • Peg8 spacer improves solubility and flexibility in conjugates.
  • Carboxylic acid functionality enables amide coupling and derivatization.
  • High purity (approximately 98.0%) for reliable synthetic performance.
  • Molecular weight 436.49 g/mol, formula C20H36O10.
  • Available in small-scale packs (100 mg) and larger sizes for scale-up.
  • Store at -20°C under inert atmosphere; in solvent, follow recommended cold storage times.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.