Learn More
Medchemexpress LLC Propargyl-PEG8-acid | 2055014-94-1 | 98.0% | 5 G
SDP
Catalog No. 5000338329
Encompass_Preferred
Propargyl-PEG8-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a cleavable ADC linker for antibody-drug conjugates (ADCs), applicable in bacterial infections caused by Gram-negative bacteria. This product is also a click chemistry reagent, featuring an Alkyne group that participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules.
  • Click chemistry
  • Alkyne applications
  • ADC synthesis
  • PROTAC synthesis
  • Supports research in infection and cancer
  • Used in cancer targeted therapy and immunotherapy
  • Relevant for bacterial studies
Catalog No. 50-003-38329 Supplier Medchemexpress LLC Supplier No. HY1303795G
Propargyl-PEG8-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a cleavable ADC linker for antibody-drug conjugates (ADCs), applicable in bacterial infections caused by Gram-negative bacteria. This product is also a click chemistry reagent, featuring an Alkyne group that participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules.
  • Click chemistry
  • Alkyne applications
  • ADC synthesis
  • PROTAC synthesis
  • Supports research in infection and cancer
  • Used in cancer targeted therapy and immunotherapy
  • Relevant for bacterial studies