Learn More
Medchemexpress LLC Propargyl-PEG8-Boc | 2055014-96-3 | 1 G
SDP
Catalog No. 5000339064
Encompass_Preferred
Propargyl-PEG8-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups. This compound is instrumental in developing PROTACs, which are designed to selectively degrade target proteins by utilizing the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an Alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Interacts with Azide groups
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-39064 Supplier Medchemexpress LLC Supplier No. HY1303751G
Propargyl-PEG8-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups. This compound is instrumental in developing PROTACs, which are designed to selectively degrade target proteins by utilizing the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an Alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Interacts with Azide groups
  • Facilitates selective degradation of target proteins