Learn More
Medchemexpress LLC Propargyl-PEG8-Boc | 2055014-96-3 | 100 MG
SDP
Catalog No. 5000339232
Encompass_Preferred
Propargyl-PEG8-Boc is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This click chemistry reagent features an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing Azide groups. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Reacts with azide-containing molecules
Catalog No. 50-003-39232 Supplier Medchemexpress LLC Supplier No. HY130375100MG
Propargyl-PEG8-Boc is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This click chemistry reagent features an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing Azide groups. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Reacts with azide-containing molecules