Learn More
Medchemexpress LLC Propargyl-PEG8-Boc | 2055014-96-3 | 250 MG
SDP
Catalog No. 5000341491
Encompass_Preferred
Propargyl-PEG8-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-41491 Supplier Medchemexpress LLC Supplier No. 250MGHY130375
Propargyl-PEG8-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins