Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG8-bromide | 2055046-25-6 | 471.38 | C19H35BrO8 | 1 G
SDP

Catalog No. 5000202826
Encompass_Preferred

Propargyl-PEG8-bromide is a PEG-based heterobifunctional linker containing a terminal propargyl (alkyne) group and a bromide leaving group. It is intended for use in ADC and PROTAC linker construction and for click-chemistry bioconjugation.

  • Heterobifunctional PEG linker with terminal alkyne and bromide.
  • Molecular formula C19H35BrO8 and molecular weight 471.38.
  • Used in ADC and PROTAC linker synthesis and alkyne-azide click chemistry.
  • Supplied in small-scale packages, including 1 g.
  • CAS number 2055046-25-6 for unambiguous identification.

Catalog No. 50-002-02826 Supplier Medchemexpress LLC Supplier No. HY1303771G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG8-bromide is a PEG-based heterobifunctional linker containing a terminal propargyl (alkyne) group and a bromide leaving group. It is intended for use in ADC and PROTAC linker construction and for click-chemistry bioconjugation.

  • Heterobifunctional PEG linker with terminal alkyne and bromide.
  • Molecular formula C19H35BrO8 and molecular weight 471.38.
  • Used in ADC and PROTAC linker synthesis and alkyne-azide click chemistry.
  • Supplied in small-scale packages, including 1 g.
  • CAS number 2055046-25-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.